Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031533.1 | NP_001026704.1 | 3 - 4 | 435 | 74 | |
GenBank mRNA | AJ721054.1 | - | 3 - 4 | 435 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |