Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031540.1 | NP_001026711.1 | 5 - 6 | 493 | 77 | |
XM_025149752.1 | XP_025005520.1 | 5 - 6 | 409 | 77 | ||
GenBank mRNA | AJ720771.1 | - | 5 - 6 | 493 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |