Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031541.2 | NP_001026712.2 | 2 - 3 | 241 | 104 | |
GenBank mRNA | AJ720426.1 | - | 2 - 3 | 212 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |