Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031543.1 | NP_001026714.1 | 1 | 45 | 70 | |
XM_025150824.1 | XP_025006592.1 | 1 | 45 | 70 | ||
XM_025150825.1 | XP_025006593.1 | 1 | 45 | 70 | ||
XM_025150826.1 | XP_025006594.1 | 1 | 45 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |