Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031562.1 | NP_001026733.1 | 4 - 5 | 514 | 119 | |
GenBank mRNA | AJ851716.1 | - | 4 - 5 | 514 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |