Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031562.1 | NP_001026733.1 | 6 - 7 | 784 | 66 | |
GenBank mRNA | AJ851716.1 | - | 6 - 7 | 784 | 66 | |
CR354158.1 | - | 1 - 2 | 72 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |