Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: ZC3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031585.2 | NP_001026756.1 | 10 - 11 | 2586 | 140 | |
XM_015283175.2 | XP_015138661.1 | 10 - 11 | 2652 | 140 | ||
GenBank mRNA | AJ851559.1 | - | 10 - 11 | 2587 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |