Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031595.1 | NP_001026766.1 | 3 - 4 | 320 | 60 | |
GenBank mRNA | AJ719672.1 | - | 3 - 4 | 320 | 60 | |
BX934361.2 | - | 3 - 4 | 278 | 60 | ||
CR405724.1 | - | 3 - 4 | 272 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |