Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030336.1 | NP_001025507.1 | 2 - 3 | 645 | 69 | |
XM_015282366.2 | XP_015137852.1 | 2 - 3 | 1817 | 69 | ||
GenBank mRNA | AJ237599.1 | - | 2 - 3 | 645 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |