Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278153.1 | NP_001265082.1 | 2 - 3 | 247 | 95 | |
XM_015294751.2 | XP_015150237.1 | 2 - 3 | 245 | 95 | ||
GenBank mRNA | AJ719523.1 | - | 2 - 3 | 247 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |