Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030652.1 | NP_001025823.1 | 4 - 5 | 644 | 96 | |
XM_015275227.2 | XP_015130713.1 | 4 - 5 | 662 | 96 | ||
GenBank mRNA | AJ719634.1 | - | 4 - 5 | 644 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |