Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030654.2 | NP_001025825.2 | 11 - 12 | 1096 | 76 | |
XM_015275241.2 | XP_015130727.1 | 10 - 11 | 1284 | 76 | ||
XM_025155434.1 | XP_025011202.1 | 9 - 10 | 1317 | 76 | ||
XM_025155435.1 | XP_025011203.1 | 10 - 11 | 4286 | 76 | ||
XM_025155436.1 | XP_025011204.1 | 11 - 12 | 4855 | 76 | ||
XM_025155437.1 | XP_025011205.1 | 10 - 11 | 1234 | 76 | ||
GenBank mRNA | AJ446747.1 | - | 3 - 4 | 299 | 76 | |
AJ851383.1 | - | 11 - 12 | 1096 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |