Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030663.1 | NP_001025834.1 | 5 - 6 | 717 | 82 | |
XM_015275297.2 | XP_015130783.1 | 5 - 6 | 720 | 82 | ||
XM_015275298.2 | XP_015130784.1 | 5 - 6 | 797 | 82 | ||
XM_025155462.1 | XP_025011230.1 | 6 - 7 | 810 | 82 | ||
XM_025155463.1 | XP_025011231.1 | 5 - 6 | 694 | 82 | ||
XM_025155464.1 | XP_025011232.1 | 5 - 6 | 707 | 82 | ||
GenBank mRNA | AJ851380.1 | - | 5 - 6 | 717 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |