Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM27.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030671.2 | NP_001025842.2 | 4 - 5 | 746 | 103 | |
XM_015294956.2 | XP_015150442.1 | 3 - 4 | 1027 | 103 | ||
XM_015294957.2 | XP_015150443.1 | 4 - 5 | 1120 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |