Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030682.1 | NP_001025853.1 | 3 - 4 | 503 | 102 | |
XM_025156062.1 | XP_025011830.1 | 3 - 4 | 495 | 102 | ||
GenBank mRNA | AJ721097.1 | - | 3 - 4 | 503 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |