Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030688.1 | NP_001025859.1 | 5 - 6 | 656 | 83 | |
XM_015279473.2 | XP_015134959.1 | 4 - 5 | 708 | 83 | ||
XM_015279474.2 | XP_015134960.1 | 5 - 6 | 725 | 83 | ||
GenBank mRNA | AJ851562.1 | - | 5 - 6 | 656 | 83 | |
CR338692.1 | - | 644 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |