Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030700.1 | NP_001025871.1 | 6 - 7 | 967 | 71 | |
GenBank mRNA | AJ720296.1 | - | 6 - 7 | 967 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |