Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030713.1 | NP_001025884.1 | 5 - 6 | 479 | 108 | |
GenBank mRNA | AJ720622.1 | - | 5 - 6 | 479 | 108 | |
CR391671.1 | - | 3 - 4 | 205 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |