Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030716.2 | NP_001025887.2 | 10 - 11 | 1093 | 93 | |
XM_015295755.2 | XP_015151241.1 | 10 - 11 | 1194 | 93 | ||
GenBank mRNA | AJ721005.1 | - | 10 - 11 | 1093 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |