Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAMPTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030728.1 | NP_001025899.1 | 10 - 11 | 1427 | 148 | |
XM_015275716.2 | XP_015131202.1 | 11 - 12 | 1698 | 148 | ||
GenBank mRNA | AY946242.1 | - | 10 - 11 | 1427 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |