Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030730.1 | NP_001025901.1 | 2 - 3 | 528 | 109 | |
XM_015275728.2 | XP_015131214.1 | 2 - 3 | 987 | 109 | ||
XM_015275729.2 | XP_015131215.1 | 2 - 3 | 492 | 109 | ||
GenBank mRNA | AJ719820.1 | - | 2 - 3 | 528 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |