Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030736.1 | NP_001025907.1 | 6 - 7 | 1036 | 118 | |
XM_015281195.1 | XP_015136681.1 | 5 - 6 | 1001 | 118 | ||
GenBank mRNA | AJ720098.1 | - | 6 - 7 | 1036 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |