Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030894.2 | NP_001026065.1 | 3 - 4 | 408 | 87 | |
XM_015297648.2 | XP_015153134.1 | 4 - 5 | 555 | 87 | ||
GenBank mRNA | AJ719961.1 | - | 3 - 4 | 409 | 87 | |
BX930386.1 | - | 3 - 4 | 391 | 87 | ||
BX930954.1 | - | 4 - 5 | 485 | 87 | ||
BX932588.1 | - | 3 - 4 | 392 | 87 | ||
CR389031.1 | - | 3 - 4 | 389 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |