Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305188.1 | NP_001292117.1 | 4 - 5 | 420 | 76 | |
GenBank mRNA | AJ720928.1 | - | 4 - 5 | 420 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |