Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030906.1 | NP_001026077.1 | 1 - 2 | 161 | 71 | |
XM_015297675.2 | XP_015153161.1 | 1 - 2 | 195 | 71 | ||
XM_015297677.2 | XP_015153163.1 | 1 - 2 | 195 | 71 | ||
XM_015297678.2 | XP_015153164.1 | 1 - 2 | 265 | 71 | ||
XM_015297679.2 | XP_015153165.1 | 1 - 2 | 195 | 71 | ||
XM_025142998.1 | XP_024998766.1 | 1 - 2 | 195 | 71 | ||
GenBank mRNA | AJ719320.1 | - | 1 - 2 | 161 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |