Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030951.1 | NP_001026122.1 | 3 - 4 | 474 | 103 | |
GenBank mRNA | AJ719788.1 | - | 3 - 4 | 474 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |