Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYD88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030962.4 | NP_001026133.2 | 2 - 3 | 480 | 107 | |
GenBank mRNA | AJ851640.1 | - | 2 - 3 | 483 | 107 | |
EF011109.1 | - | 2 - 3 | 474 | 107 | ||
EF544486.1 | - | 2 - 3 | 467 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |