Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030986.1 | NP_001026157.1 | 7 - 8 | 748 | 106 | |
XM_015281467.1 | XP_015136953.1 | 6 - 7 | 782 | 106 | ||
XM_015281468.2 | XP_015136954.1 | 6 - 7 | 652 | 106 | ||
XM_025147204.1 | XP_025002972.1 | 7 - 8 | 778 | 106 | ||
GenBank mRNA | AJ719665.1 | - | 7 - 8 | 809 | 106 | |
AJ720825.1 | - | 7 - 8 | 748 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |