Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031001.1 | NP_001026172.1 | 2 - 3 | 224 | 91 | |
XM_015282171.1 | XP_015137657.1 | 1 - 2 | 2183 | 91 | ||
GenBank mRNA | AB288388.1 | - | 1 - 2 | 160 | 91 | |
AB288389.1 | - | 1 - 2 | 160 | 91 | ||
AB288390.1 | - | 1 - 2 | 160 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |