Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031276.1 | NP_001026447.1 | 30 - 31 | 4920 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |