Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031276.1 | NP_001026447.1 | 33 - 34 | 5265 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |