Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031288.1 | NP_001026459.1 | 1 - 2 | 333 | 52 | |
GenBank mRNA | AJ720832.1 | - | 1 - 2 | 333 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |