Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031303.2 | NP_001026474.2 | 1 - 2 | 357 | 138 | |
GenBank mRNA | AJ450241.1 | - | 1 - 2 | 355 | 138 | |
AJ851379.1 | - | 1 - 2 | 278 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |