Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031310.1 | NP_001026481.1 | 3 - 4 | 308 | 93 | |
GenBank mRNA | AJ720946.1 | - | 3 - 4 | 308 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |