Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031312.1 | NP_001026483.1 | 3 - 4 | 317 | 73 | |
XM_015276923.2 | XP_015132409.1 | 3 - 4 | 355 | 73 | ||
GenBank mRNA | AJ720550.1 | - | 3 - 4 | 296 | 73 | |
AJ721011.1 | - | 3 - 4 | 317 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |