Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031313.1 | NP_001026484.1 | 1 - 2 | 175 | 129 | |
XM_015291390.2 | XP_015146876.1 | 1 - 2 | 186 | 129 | ||
GenBank mRNA | AJ851823.1 | - | 1 - 2 | 175 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |