Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145746.1 | XP_025001514.1 | 18 - 19 | 3220 | 87 | |
XM_025145747.1 | XP_025001515.1 | 18 - 19 | 3217 | 87 | ||
XM_025145748.1 | XP_025001516.1 | 15 - 16 | 2956 | 87 |
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ851711.1 | - | 18 - 19 | 3134 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |