Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031375.3 | NP_001026546.2 | 7 - 8 | 1216 | 119 | |
XM_015285445.2 | XP_015140931.1 | 6 - 7 | 882 | 119 | ||
GenBank mRNA | AJ720001.1 | - | 7 - 8 | 1225 | 119 | |
CR523286.1 | - | 5 - 6 | 679 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |