Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM76A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031377.2 | NP_001026548.1 | 6 - 7 | 635 | 150 | |
GenBank mRNA | AJ720569.1 | - | 6 - 7 | 614 | 150 | |
CR389356.1 | - | 1 - 2 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |