Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNSTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031393.1 | NP_001026564.1 | 6 - 7 | 624 | 99 | |
GenBank mRNA | AJ721023.1 | - | 6 - 7 | 624 | 99 | |
CR352610.1 | - | 5 - 6 | 478 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |