Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031409.1 | NP_001026580.1 | 10 - 11 | 1091 | 92 | |
XM_015293130.2 | XP_015148616.1 | 11 - 12 | 1336 | 92 | ||
XM_015293131.2 | XP_015148617.1 | 11 - 12 | 1208 | 92 | ||
GenBank mRNA | AJ868233.1 | - | 10 - 11 | 1091 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |