Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031409.1 | NP_001026580.1 | 9 - 10 | 1004 | 73 | |
XM_015293130.2 | XP_015148616.1 | 10 - 11 | 1249 | 73 | ||
XM_015293131.2 | XP_015148617.1 | 10 - 11 | 1121 | 73 | ||
XM_015293132.2 | XP_015148618.1 | 10 - 11 | 1251 | 73 | ||
GenBank mRNA | AJ868233.1 | - | 9 - 10 | 1004 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |