Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031773.1 | NP_001026943.1 | 2 - 3 | 541 | 92 | |
XM_025153331.1 | XP_025009099.1 | 2 - 3 | 249 | 92 | ||
GenBank mRNA | AY971655.1 | - | 2 - 3 | 541 | 92 | |
BX933977.2 | - | 2 - 3 | 444 | 92 | ||
BX934902.1 | - | 1 - 2 | 144 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |