Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033869.2 | NP_001029041.1 | 6 - 7 | 669 | 107 | |
XM_015294017.2 | XP_015149503.1 | 6 - 7 | 747 | 107 | ||
XM_025154858.1 | XP_025010626.1 | 6 - 7 | 590 | 107 | ||
GenBank mRNA | CR390115.1 | - | 6 - 7 | 670 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |