Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034814.2 | NP_001029986.2 | 15 - 16 | 1463 | 132 | |
XM_025154152.1 | XP_025009920.1 | 22 - 23 | 5747 | 132 | ||
GenBank mRNA | AB205527.1 | - | 15 - 16 | 1489 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |