Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034816.3 | NP_001029988.2 | 4 - 5 | 855 | 149 | |
GenBank mRNA | AJ005071.1 | - | 2 - 3 | 525 | 149 | |
AM403065.1 | - | 4 - 5 | 825 | 149 | ||
AM403066.1 | - | 4 - 5 | 825 | 149 | ||
AM403067.1 | - | 4 - 5 | 825 | 149 | ||
AM403068.1 | - | 4 - 5 | 825 | 149 | ||
AM403069.1 | - | 4 - 5 | 825 | 149 | ||
AM403070.1 | - | 4 - 5 | 825 | 149 | ||
AM403071.1 | - | 4 - 5 | 825 | 149 | ||
AM403072.1 | - | 4 - 5 | 825 | 149 | ||
AM403073.1 | - | 4 - 5 | 825 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |