Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034817.2 | NP_001029989.1 | 8 - 9 | 965 | 135 | |
GenBank mRNA | AY847576.1 | - | 8 - 9 | 966 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |