Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034818.1 | NP_001029990.1 | 2 - 3 | 353 | 80 | |
XM_015284525.2 | XP_015140011.1 | 3 - 4 | 531 | 80 | ||
XM_015284527.2 | XP_015140013.1 | 3 - 4 | 528 | 80 | ||
XM_015284528.2 | XP_015140014.1 | 2 - 3 | 2205 | 80 | ||
GenBank mRNA | CR353371.1 | - | 2 - 3 | 353 | 80 | |
DQ127227.1 | - | 2 - 3 | 353 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |