Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039255.1 | NP_001034344.1 | 7 - 8 | 870 | 77 | |
GenBank mRNA | AJ720244.1 | - | 7 - 8 | 870 | 77 | |
BX932666.2 | - | 7 - 8 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |