Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EWSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039267.2 | NP_001034356.2 | 3 - 4 | 136 | 69 | |
XM_015294853.2 | XP_015150339.1 | 3 - 4 | 124 | 69 | ||
XM_015294854.2 | XP_015150340.1 | 3 - 4 | 124 | 69 | ||
XM_015294855.2 | XP_015150341.1 | 3 - 4 | 142 | 69 | ||
XM_015294856.2 | XP_015150342.1 | 3 - 4 | 142 | 69 | ||
GenBank mRNA | AJ719366.1 | - | 3 - 4 | 136 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |